SY3H3D67BA07 Synthonix Corporation

Dimethyl Methoxymalonate

  • MDL number MFCD00009847
  • Cas Number : 5018-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41479.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41479.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart