SY3H3D67BA20 Synthonix Corporation

3,3'-Disulfanediylbis(propan-1-ol)

  • MDL number MFCD01630941
  • Cas Number : 30453-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41614.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41614.html
pictograms GHS07
hstatements H227, H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart