SY3H3D67BA26 Synthonix Corporation

1,2-Dibromo-5-chloro-3-fluorobenzene

  • MDL number MFCD00143441
  • Cas Number : 208186-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41629.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41629.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart