SY3H3D67BA4F Synthonix Corporation

2,5-Dibromothiophene-3,4-dicarboxylic acid

  • MDL number MFCD08695787
  • Cas Number : 190723-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41769.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41769.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart