SY3H3D67BA54 Synthonix Corporation

1,3-Dibromo-2-iodobenzene

  • Cas Number : 19821-80-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41775.html

Safety Information

MSDS https://synthonix.com/41775.html
Availability details for
Quantity
  • Availability
  • Add to Cart