SY3H3D67BA56 Synthonix Corporation

3,4-Dichlorofluorobenzene

  • MDL number MFCD00018119
  • Cas Number : 1435-49-0
  • Molecular Weight : 164.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41797.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41797.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart