SY3H3D67BA5D Synthonix Corporation

1,2-Difluoro-4-iodobenzene

  • MDL number MFCD00044630
  • Cas Number : 64248-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Yellow - Red-brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41813.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41813.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart