SY3H3D67BA5E Synthonix Corporation

1,3-Difluoro-2-iodobenzene

  • MDL number MFCD04039247
  • Cas Number : 13697-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Liquid / Colour: White - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41814.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41814.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart