SY3H3D67BA64 Synthonix Corporation

2,7-Dibromo-5H-pyrrolo[2,3-b]pyrazine

  • MDL number MFCD11109803
  • Cas Number : 1209703-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41861.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41861.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart