SY3H3D67BA71 Synthonix Corporation

1,3-Dibromo-2-nitrobenzene

  • Cas Number : 13402-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41900.html

Safety Information

MSDS https://synthonix.com/41900.html
Availability details for
Quantity
  • Availability
  • Add to Cart