SY3H3D67BA78 Synthonix Corporation

1,3-Dichloro-2-nitrobenzene

  • MDL number MFCD00456795
  • Cas Number : 601-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Pale yellow - Yellow or Green
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41929.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41929.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart