SY3H3D67BA9C Synthonix Corporation

2,4-Dichloro-6,7-dihydrothieno[3,2-d]pyrimidine

  • Cas Number : 74901-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42071.html

Safety Information

MSDS https://synthonix.com/42071.html
Availability details for
Quantity
  • Availability
  • Add to Cart