SY3H3D67BAA3 Synthonix Corporation

2,6-Difluoro-3-nitroaniline

  • Cas Number : 25892-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42112.html

Safety Information

MSDS https://synthonix.com/42112.html
Availability details for
Quantity
  • Availability
  • Add to Cart