SY3H3D67BAB2 Synthonix Corporation

2,3-Dichloro-4-fluoroaniline

  • MDL number MFCD08706409
  • Cas Number : 36556-52-2
  • Molecular Weight : 180.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown to Brown Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/42221.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42221.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart