SY3H3D67BAC1 Synthonix Corporation

2,4-Dichlorobenzene-1,3-diol

  • MDL number MFCD13152136
  • Cas Number : 16606-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42318.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42318.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart