SY3H3D67BACD Synthonix Corporation

3,5-Dibromoaniline

  • MDL number MFCD00047841
  • Cas Number : 626-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to brown to black or gray powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42514.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42514.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart