SY3H3D67BAD5 Synthonix Corporation

2,5-Dichloro-3-methylpyridine

  • MDL number MFCD03840754
  • Cas Number : 59782-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42560.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42560.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart