SY3H3D67BAE8 Synthonix Corporation

2,3-Dichloro-6-methoxypyridine

  • Cas Number : 83732-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42661.html

Safety Information

MSDS https://synthonix.com/42661.html
Availability details for
Quantity
  • Availability
  • Add to Cart