SY3H3D67BAEA Synthonix Corporation

O-(3,5-Dichlorophenyl)hydroxylamine

  • MDL number MFCD09029771
  • Cas Number : 99907-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42663.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42663.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart