SY3H3D67BAFA Synthonix Corporation

2,5-Dichloro-4,6-dimethylpyrimidine

  • Cas Number : 19573-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42747.html

Safety Information

MSDS https://synthonix.com/42747.html
Availability details for
Quantity
  • Availability
  • Add to Cart