SY3H3D67BAFD Synthonix Corporation

3,5-Dichloro-4-methylpyridin-2-amine

  • MDL number MFCD00129031
  • Cas Number : 31430-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow to Yellow red or Light-gray or Pale-purple
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42750.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42750.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart