SY3H3D67BB06 Synthonix Corporation

3,4-Difluorobenzene-1,2-diamine

  • MDL number MFCD00153128
  • Cas Number : 153505-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42766.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42766.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart