SY3H3D67BB11 Synthonix Corporation

(6-(Difluoromethoxy)pyridin-3-yl)boronic acid

  • Cas Number : 1354290-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42867.html

Safety Information

MSDS https://synthonix.com/42867.html
Availability details for
Quantity
  • Availability
  • Add to Cart