SY3H3D67BB15 Synthonix Corporation

4,6-Dichloro-2-ethylpyrimidin-5-amine

  • MDL number MFCD11219203
  • Cas Number : 6237-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43088.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43088.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart