SY3H3D67BB27 Synthonix Corporation

3,5-Dimethoxypyridazine

  • Cas Number : 2096-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43215.html

Safety Information

MSDS https://synthonix.com/43215.html
Availability details for
Quantity
  • Availability
  • Add to Cart