SY3H3D67BB34 Synthonix Corporation

3,4-Dihydro-2H-pyran-6-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 31518-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43311.html

Safety Information

MSDS https://synthonix.com/43311.html
Availability details for
Quantity
  • Availability
  • Add to Cart