SY3H3D67BB3F Synthonix Corporation

2,5-Dimethoxypyrimidin-4-amine

  • MDL number MFCD20486065
  • Cas Number : 6960-17-4
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-dimethoxy-4-pyrimidinamine
Inchi Code 1S/C6H9N3O2/c1-10-4-3-8-6(11-2)9-5(4)7/h3H,1-2H3,(H2,7,8,9)
Inchi Key XOJPOIGPYBYDJF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/43359.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43359.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart