SY3H3D67BB47 Synthonix Corporation

1,2-Dibromo-4,5-difluoro-3-nitrobenzene

  • MDL number MFCD11226312
  • Cas Number : 1481-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43415.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43415.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart