SY3H3D67BB48 Synthonix Corporation

4,7-Dibromo-5,6-difluorobenzo[c][1,2,5]thiadiazole

  • MDL number MFCD28369502
  • Cas Number : 1295502-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Light-gray - Pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43419.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/43419.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart