SY3H3D67BB58 Synthonix Corporation

2,6-Diaminophenol hydrochloride

  • Cas Number : 1808091-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43538.html

Safety Information

MSDS https://synthonix.com/43538.html
Availability details for
Quantity
  • Availability
  • Add to Cart