SY3H3D67BB67 Synthonix Corporation

2,6-Dimethylpyridin-3-amine

  • MDL number MFCD01646409
  • Cas Number : 3430-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43608.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43608.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart