SY3H3D67BB6E Synthonix Corporation

N,6-dimethylpyridin-2-amine

  • MDL number MFCD10697830
  • Cas Number : 97986-08-8
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Liquid to Sticky Oil to Semi-Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/43615.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43615.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart