SY3H3D67BB79 Synthonix Corporation

3,4-Dimethylcyclopent-2-enone

  • MDL number MFCD09701445
  • Cas Number : 30434-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43712.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43712.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart