SY3H3D67BB84 Synthonix Corporation

Dimethyl 2-bromopentanedioate

  • MDL number MFCD00043630
  • Cas Number : 760-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43785.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43785.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart