SY3H3D67BB8B Synthonix Corporation

1-(1,3-Dimethyl-1H-pyrazol-5-yl)-N-methylmethanamine

  • MDL number No MDL found
  • Cas Number : 852227-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/43837.html

Safety Information

MSDS https://synthonix.com/43837.html
Availability details for
Quantity
  • Availability
  • Add to Cart