SY3H3D67BB9A Synthonix Corporation

2,6-Dichloro-3-nitrobenzonitrile

  • MDL number MFCD00051513
  • Cas Number : 5866-98-8
  • Molecular Weight : 217.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43973.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43973.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart