SY3H3D67BBA4 Synthonix Corporation

1,2-Dichloro-5-nitro-3-(trifluoromethyl)benzene

  • Cas Number : 400-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/44004.html

Safety Information

MSDS https://synthonix.com/44004.html
Availability details for
Quantity
  • Availability
  • Add to Cart