SY3H3D67BBB6 Synthonix Corporation

2,3-Dichlorobenzonitrile

  • MDL number MFCD00016372
  • Cas Number : 6574-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Pale yellow green
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44115.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44115.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart