SY3H3D67BBBE Synthonix Corporation

2,5-Difluoro-4-nitrobenzoic acid

  • MDL number MFCD03425705
  • Cas Number : 116465-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to yellow-brown or gray solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44172.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44172.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart