SY3H3D67BBC5 Synthonix Corporation

3,6-Dichlorobenzo[d]isoxazole

  • Cas Number : 16263-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44183.html

Safety Information

MSDS https://synthonix.com/44183.html
Availability details for
Quantity
  • Availability
  • Add to Cart