SY3H3D67BBCA Synthonix Corporation

2,4-Dichloro-5-fluorobenzoic acid

  • MDL number MFCD00075281
  • Cas Number : 86522-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44205.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44205.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart