SY3H3D67BBCD Synthonix Corporation

2,6-Difluoro-3-iodobenzoic acid

  • MDL number MFCD11040446
  • Cas Number : 229178-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44220.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44220.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart