SY3H3D67BBD3 Synthonix Corporation

1,4-Dibromo-2-(trifluoromethoxy)benzene

  • Cas Number : 175278-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44228.html

Safety Information

MSDS https://synthonix.com/44228.html
Availability details for
Quantity
  • Availability
  • Add to Cart