SY3H3D67BBEC Synthonix Corporation

1,5-Difluoro-3-methyl-2,4-dinitrobenzene

  • MDL number MFCD28099117
  • Cas Number : 1616526-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44376.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44376.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart