SY3H3D67BBF7 Synthonix Corporation

2,5-Dibromobenzoic acid

  • MDL number MFCD00016494
  • Cas Number : 610-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44563.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44563.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart