SY3H3D67BBF8 Synthonix Corporation

5,6-Dibromobenzo[d][1,3]dioxole

  • MDL number MFCD00094476
  • Cas Number : 5279-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44564.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44564.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart