SY3H3D67BBFF Synthonix Corporation

3,4-Difluoro-5-hydroxybenzaldehyde

  • Cas Number : 887584-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44609.html

Safety Information

MSDS https://synthonix.com/44609.html
Availability details for
Quantity
  • Availability
  • Add to Cart