SY3H3D67BC04 Synthonix Corporation

2,6-Difluoro-4-hydroxybenzoic acid

  • Cas Number : 214917-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/44625.html

Safety Information

MSDS https://synthonix.com/44625.html
Availability details for
Quantity
  • Availability
  • Add to Cart