SY3H3D67BC10 Synthonix Corporation

1,2-Dibromo-4-fluoro-5-methylbenzene

  • MDL number MFCD00143234
  • Cas Number : 202982-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44694.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44694.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart