SY3H3D67BC18 Synthonix Corporation

5,6-Dichloro-1H-benzo[d]imidazol-2-amine

  • Cas Number : 18672-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/44860.html

Safety Information

MSDS https://synthonix.com/44860.html
Availability details for
Quantity
  • Availability
  • Add to Cart